Learn More
Bismuth(III) selenide, Vacuum Deposition Grade, 99.995% (metals basis), Thermo Scientific Chemicals
Description
In semiconductor research, multilayer thin-film, magnetoresistant film.Bismuth(III) selenide is used as a semiconductor and thermoelectric material. It is also used in semiconductor research. Further, it finds applications magnetoresistant film and semiconductor research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 710°C |
Color | Gray |
Physical Form | Pieces |
Quantity | 50 g |
UN Number | UN3283 |
Merck Index | 14,1278 |
Solubility Information | Insoluble in water; Decomposes when heated in air; Decomposes in nitric acid and aqua regiaSoluble in strong acids; Insoluble in water and organic solvents. |
Formula Weight | 654.84 |
Percent Purity | 99.999% |
Density | 6.82 g/mL |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.