missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(Iii) Subnitrate, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
50 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 250 g |
Linear Formula | Bi5O(OH)9(NO3)4 |
UN Number | 1477 |
Formula Weight | 1461.99g/mol |
Grade | puriss. |
Concentration or Composition (by Analyte or Components) | 71 to 74% Bi basis (calc. on dried substance) |
Packaging | Plastic Bottle |
Chemical Name or Material | Bismuth(Iii) Subnitrate, Meets analytical specification of USP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'