missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(Iii) Subnitrate, Honeywell Fluka™
BIOPHARMA

Product Code. p-7071756 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
50 kg
Unit Size:
1kg
250g
50kg
This item is not returnable. View return policy

Product Code. 15690970

Brand: Honeywell-Fluka 10334250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of DAB, USP, 71-74% Bi basis (calc. on dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-85-4
MDL Number MFCD00064844

Specifications

Quantity 250 g
Linear Formula Bi5O(OH)9(NO3)4
UN Number 1477
Formula Weight 1461.99g/mol
Grade puriss.
Concentration or Composition (by Analyte or Components) 71 to 74% Bi basis (calc. on dried substance)
Packaging Plastic Bottle
Chemical Name or Material Bismuth(Iii) Subnitrate, Meets analytical specification of USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.