missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) tert-pentyloxide, 97+%, Thermo Scientific Chemicals

Product Code. 11339567
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124687-44-1
MDL Number MFCD00151666
Synonym Bismuth(III) tert-amylate; Bismuth(III) tert-pentoxide

Specifications

Melting Point 180°C (decomposition)
Color Gray
Boiling Point 63°C (at 10-4mmHg)(sublimation)
Linear Formula Bi(OC5H11)3
Quantity 1 g
UN Number UN3181
Formula Weight 470.41
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Bismuth(III) tert-pentyloxide
compliance-icons compliance-icons
Product Identifier
  • Bismuth(III) tert-pentyloxide
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'