missing translation for 'onlineSavingsMsg'
Learn More

Bismuth needles, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11364228
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Form: Approx. 1.5-3.0mm diameter; Various lengths

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Color Gray
Appearance Gray needles
Assay Percent Range 99.99% (Metals basis)
Quantity 100 g
Odor Odorless
Form Needle
Chemical Name or Material Bismuth

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'