missing translation for 'onlineSavingsMsg'
Learn More
Bismuth, plasma standard solution, Specpure™ Bi 10,000μg/mL, Thermo Scientific Chemicals Product Code.: 11352308

Bismuth, plasma standard solution, Specpure™ Bi 10,000μg/mL, Thermo Scientific Chemicals

Product Code. 11352308
50 mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11352308

Brand: Thermo Scientific Alfa Aesar 014388.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00134033

Specifications

Quantity 50 mL
UN Number UN3264
Physical Form Liquid
Concentration Bi in 5% HNO3
Grade Specpure™
Chemical Name or Material Bismuth plasma standard solution, Bi 10000μg/mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1544570 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number014388.AP,014388.AD,014388.AE
Lot Number1523468 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number014388.AP,014388.AD,014388.AE
Lot Number1517334 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number014388.AP,014388.AD,014388.AE
Lot Number1493423 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number014388.AP,014388.AD,014388.AE
Lot Number1481520 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number014388.AP,014388.AD,014388.AE
compliance-icons
Product Identifier
  • Bismuth, plasma standard solution, Specpure, Bi 10,000µg/ml
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth, plasma standard solution, Specpure™ Bi 10,000μg/mL, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.