missing translation for 'onlineSavingsMsg'
Learn More

Bismuth powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11351158
Click to view available options
Quantity:
100 g
500 g
2 kg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

UN Number UN3089
Quantity 500 g
Percent Purity 99.5%
Mesh Size −100 Mesh
Odor Odorless
Weight Apparent density: 3.8g/cm3 to 4.6g/cm3
Form Powder
Assay (metals basis)
Chemical Name or Material Bismuth powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'