Learn More
Bismuth powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
91.40 € - 506.00 €
Chemical Identifiers
CAS | 7440-69-9 |
---|---|
Molecular Formula | Bi |
Molecular Weight (g/mol) | 208.98 |
MDL Number | MFCD00134033 |
InChI Key | JCXGWMGPZLAOME-UHFFFAOYSA-N |
Synonym | bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod |
PubChem CID | 5359367 |
ChEBI | CHEBI:33301 |
IUPAC Name | bismuth |
SMILES | [Bi] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316867
|
Thermo Scientific Alfa Aesar
010766.06 |
5 g |
91.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326867
|
Thermo Scientific Alfa Aesar
010766.14 |
25 g |
206.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306867
|
Thermo Scientific Alfa Aesar
010766.22 |
100 g |
506.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-69-9 | |
208.98 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
5359367 | |
bismuth |
Bi | |
MFCD00134033 | |
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod | |
CHEBI:33301 | |
[Bi] |
Specifications
Gray | |
Gray powder | |
99.999% (Metals basis) | |
MFCD00134033 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
bismuth | |
5359367 | |
-200 Mesh | |
Powder |
7440-69-9 | |
9.79g/cm3 | |
Bi | |
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod | |
[Bi] | |
208.98 | |
CHEBI:33301 | |
Odorless | |
Bismuth |
Safety and Handling
Take precautionary measures against static discharges.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : DOT Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 231-177-4
missing translation for 'rtecsNumber' : EB2600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Bismuth powder
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P370+P378-In case of fire: Use to extinguish.
RUO – Research Use Only