Learn More
Bismuth Subsalicylate, MedChemExpress
MedChemExpress Bismuth Subsalicylate is a potent and orally active antacid and anti-diarrheal agent. Bismuth Subsalicylate reduces inflammation/irritation of stomach and intestinal lining through inhibition of prostaglandin synthesis in vivo. Bismuth Subsalicylate is widely used for the research of diarrheal disorders, including indigestion, diarrhoea, nausea, et al.
79.65 € - 143.10 €
Specifications
Chemical Name or Material | Bismuth Subsalicylate |
---|---|
CAS | 14882-18-9 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C7H5BiO4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16490988
|
Medchem Express
HY-B0550 |
5 g |
79.65 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16408528
|
Medchem Express
HY-B0550 |
10 g |
143.10 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bismuth Subsalicylate is a potent and orally active antacid and anti-diarrheal agent. Bismuth Subsalicylate reduces inflammation/irritation of stomach and intestinal lining through inhibition of prostaglandin synthesis in vivo. Bismuth Subsalicylate is widely used for the research of diarrheal disorders, including indigestion, diarrhoea, nausea, et al.Specifications
Bismuth Subsalicylate | |
White | |
C7H5BiO4 | |
DMSO : 1 mg/mL (2.76 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.28 mM; Need ultrasonic) | |
362.09 | |
Research | |
Neuroscience-Neuromodulation |
14882-18-9 | |
Research | |
Bismuth oxysalicylate Bismuth(III) salicylate basic | |
O=C1O[Bi](O)OC2=CC=CC=C12 | |
362.09 | |
Solid |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only