Learn More
Bismuth Subsalicylate, MedChemExpress
MedChemExpress Bismuth Subsalicylate is a potent and orally active antacid and anti-diarrheal agent. Bismuth Subsalicylate reduces inflammation/irritation of stomach and intestinal lining through inhibition of prostaglandin synthesis in vivo. Bismuth Subsalicylate is widely used for the research of diarrheal disorders, including indigestion, diarrhoea, nausea, et al.
Brand: Medchem Express HY-B0550
This item is not returnable.
View return policy
Description
Bismuth Subsalicylate is a potent and orally active antacid and anti-diarrheal agent. Bismuth Subsalicylate reduces inflammation/irritation of stomach and intestinal lining through inhibition of prostaglandin synthesis in vivo. Bismuth Subsalicylate is widely used for the research of diarrheal disorders, including indigestion, diarrhoea, nausea, et al.Specifications
Bismuth Subsalicylate | |
White | |
C7H5BiO4 | |
Bismuth oxysalicylate Bismuth(III) salicylate basic | |
O=C1O[Bi](O)OC2=CC=CC=C12 | |
362.09 | |
Solid |
14882-18-9 | |
Research | |
10 g | |
DMSO : 1 mg/mL (2.76 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.28 mM; Need ultrasonic) | |
362.09 | |
Research | |
Neuroscience-Neuromodulation |
Research purposes only