missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Tin eutectic lump, 99.95% (metals basis), Thermo Scientific Chemicals

51.15 € - 553.76 €

Specifications

Chemical Name or Material Bismuth Tin eutectic lump
Melting Point 138°C
Avg. Mol. Wt. or Mol. Wt. Range Bi:Sn; 58:42 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD00143771
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381819
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
012480.14
25 g
51.15 €
25g
Please to purchase this item. Need a web account? Register with us today!
11371819
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
012480.22
100 g
143.72 €
100g
Please to purchase this item. Need a web account? Register with us today!
11361819
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
012480.36
500 g
553.76 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bismuth Tin eutectic lump
Bi:Sn; 58:42 wt%
MFCD00143771
Lump
138°C
(metals basis)
99.95%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only