missing translation for 'onlineSavingsMsg'
Learn More

Bismuth titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11308818
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12233-34-0
Molecular Formula Bi2O11Ti4
Molecular Weight (g/mol) 785.418
MDL Number MFCD01310393
InChI Key TZQKEPWGUIJJIY-UHFFFAOYSA-N
Synonym tetraoxo-2,4,6,8,10-pentaoxa-1,11-dibisma-3,5,7,9-tetratitanaundecane-1,11-dione
PubChem CID 56846076
IUPAC Name oxo-oxobismuthanyloxy-[oxo-[oxo-(oxo(oxobismuthanyloxy)titanio)oxytitanio]oxytitanio]oxytitanium
SMILES O=[Ti](O[Ti](=O)O[Ti](=O)O[Bi]=O)O[Ti](=O)O[Bi]=O

Specifications

Physical Form Powder
Quantity 250 g
Formula Weight 785.42
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Bismuth titanium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.