missing translation for 'onlineSavingsMsg'
Learn More

Bismuth titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

99.09 € - 311.55 €

Chemical Identifiers

CAS 12233-34-0
Molecular Formula Bi2O11Ti4
Molecular Weight (g/mol) 785.418
MDL Number MFCD01310393
InChI Key TZQKEPWGUIJJIY-UHFFFAOYSA-N
Synonym tetraoxo-2,4,6,8,10-pentaoxa-1,11-dibisma-3,5,7,9-tetratitanaundecane-1,11-dione
PubChem CID 56846076
IUPAC Name oxo-oxobismuthanyloxy-[oxo-[oxo-(oxo(oxobismuthanyloxy)titanio)oxytitanio]oxytitanio]oxytitanium
SMILES O=[Ti](O[Ti](=O)O[Ti](=O)O[Bi]=O)O[Ti](=O)O[Bi]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398808
View Documents
Thermo Scientific Alfa Aesar
041669.18
50 g
99.09 €
50g
Please to purchase this item. Need a web account? Register with us today!
11308818
View Documents
Thermo Scientific Alfa Aesar
041669.30
250 g
311.55 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12233-34-0
785.418
TZQKEPWGUIJJIY-UHFFFAOYSA-N
56846076
O=[Ti](O[Ti](=O)O[Ti](=O)O[Bi]=O)O[Ti](=O)O[Bi]=O
Bi2O11Ti4
MFCD01310393
tetraoxo-2,4,6,8,10-pentaoxa-1,11-dibisma-3,5,7,9-tetratitanaundecane-1,11-dione
oxo-oxobismuthanyloxy-[oxo-[oxo-(oxo(oxobismuthanyloxy)titanio)oxytitanio]oxytitanio]oxytitanium

Specifications

12233-34-0
Bi2O11Ti4
tetraoxo-2,4,6,8,10-pentaoxa-1,11-dibisma-3,5,7,9-tetratitanaundecane-1,11-dione
O=[Ti](O[Ti](=O)O[Ti](=O)O[Bi]=O)O[Ti](=O)O[Bi]=O
785.418
785.42
Odorless
Bismuth titanium oxide
Powder
MFCD01310393
TZQKEPWGUIJJIY-UHFFFAOYSA-N
oxo-oxobismuthanyloxy-[oxo-[oxo-(oxo(oxobismuthanyloxy)titanio)oxytitanio]oxytitanio]oxytitanium
56846076
99.9%
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.