Learn More
Bismuth vanadium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
121.00 € - 427.13 €
Chemical Identifiers
CAS | 14059-33-7 |
---|---|
Molecular Formula | BiO4V |
Molecular Weight (g/mol) | 323.918 |
MDL Number | MFCD00064836 |
InChI Key | YNOWMDYNXXVJMC-UHFFFAOYSA-N |
Synonym | Bismuth vanadate |
PubChem CID | 20243764 |
IUPAC Name | bismuth;trioxido(oxo)vanadium |
SMILES | [O-][V](=O)([O-])[O-].[Bi+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333618
|
Thermo Scientific Alfa Aesar
039220.14 |
25 g | N/A | ||||||
11343618
|
Thermo Scientific Alfa Aesar
039220.22 |
100 g |
427.13 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bismuth vanadium oxide is used as a pigment due to its hiding power. It also finds use in paints, coatings, plastic and rubber products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14059-33-7 | |
323.918 | |
YNOWMDYNXXVJMC-UHFFFAOYSA-N | |
20243764 | |
[O-][V](=O)([O-])[O-].[Bi+3] |
BiO4V | |
MFCD00064836 | |
Bismuth vanadate | |
bismuth;trioxido(oxo)vanadium |
Specifications
14059-33-7 | |
Powder | |
MFCD00064836 | |
Bismuth vanadate | |
YNOWMDYNXXVJMC-UHFFFAOYSA-N | |
bismuth;trioxido(oxo)vanadium | |
20243764 | |
99.9% | |
Bismuth vanadium oxide |
Red to Yellow | |
BiO4V | |
UN3285 | |
Soluble in acids; Insoluble in water. | |
[O-][V](=O)([O-])[O-].[Bi+3] | |
323.918 | |
323 | |
(metals basis) |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
missing translation for 'einecsNumber' : 237-898-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Bismuth vanadium oxide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only