missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Black 96-Well Immuno Plates

missing translation for 'orderingAttributeHoverText'
Surface Treatment:
High Binding
MaxiSorp
Medium Binding
PolySorp
Universal Binding
Volume (Metric) Working:
280 μL
350 μL
missing translation for 'unitSize'
Pack of 50
Pack of 80
Beschreibung
Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ Black 96-Well Immuno Plates. Featuring a choice of three surface types and optimal binding, these plates minimize background in fluorescence reading.
Features:
- Plate has SBS footprint — fits standard equipment
- Flat-bottom wells
- Certified binding homogeneity/reproducibility
Choice of surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) are for absorption of hydrophobic molecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
- Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules.
Compatible with:
Compatible with lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country
Spezifikation
Spezifikation
| Binding Property | Hydrophobic |
| Format | Standard |
| Lid | Without Lid |
| Detection Method | Fluorescence |
| Array | 8 x 12 |
| Packaging | 80 Cases |
| Binding Type | Adsorption |
| Color | Black |
| Product Type | Microplate |
| Material | Polystyrene |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur