missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Black 96-Well Immuno Plates
Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with these black, 96-well immuno plates, available in a choice of three surface types.
Brand: Thermo Scientific™ 7605
181.85 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ Black 96-Well Immuno Plates. Featuring a choice of three surface types and optimal binding, these plates minimize background in fluorescence reading.
Features:
- Plate has SBS footprint — fits standard equipment
- Flat-bottom wells
- Certified binding homogeneity/reproducibility
Choice of surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) are for absorption of hydrophobic molecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
- Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules.
Ordering Information:
Availability may vary by country
Specifications
Hydrophobic | |
Without Lid | |
8 x 12 | |
Black | |
Polystyrene | |
280 μL | |
Flat Bottom (F-well) | |
50 | |
None | |
Lipids |
Standard | |
Fluorescence | |
50 Cases | |
Microplate | |
96 | |
Medium Binding | |
Fluorescence | |
10 | |
50/Cs. | |
F96 |