Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
BIOPHARMAGREENER_CHOICE
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
Brand:  Axygen™ HTF-50-CBK-HTR-S-5
 View more versions of this product
Product Code. 15561413
  • 855.00 € / Pack of 5760
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Available in non-sterile, sterile, nonfiltered, and filtered
  • Available as Clear or Conductive black polypropylene tips
  • Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 μL
Sterile
Not Autoclavable
Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Automation
Hamilton MICROLAB Nimbus
Filtered
Black
DNase, RNase and Pyrogen free
Hanging Tip Rack
5 Racks/Pk, 12 Pks/Cs.
Polypropylene