missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Black Microfiber Cloth
Click to view available options
Microfiber
Description
- Black microfiber cleaning cloth
- For scratch-free cleaning of Uvex™ safety glasses and lenses
Specifications
Specifications
Color | Black |
For Use With (Equipment) | Uvex™ Safety Spectacles |
For Use With (Application) | Scratch-Free Cleaning |
Material | Microfiber |
Disposable | Reusable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'