Learn More
Fisherbrand™ Type 1 Clear Glass Vials, with Dropper Assembly
Provides for accurate, reliable dispensing of various samples. Fisherbrand™ Type 1 Clear Glass Vials, with Dropper Assembly are manufactured from clear Type 1 neutral glass.
Brand:  Fisherbrand™ 800058
 View more versions of this product
Product Code. 14863572
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • With screw neck
  • With black caps and bulbs
  • Droppers are assembled but not fitted to vials
  • Dropper assembly comprised of clear glass bottle dropper tube fitted to a polypropylene cap with a natural rubber bulb
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
Screw
Polypropylene
43 mm
Screw Thread Vial
20 mm
5 mL
Screw
Dropper Screw Cap
40/Pk.