Learn More
BRAND™ BLAUBRAND™ Bulb Pipettes 1-Mark
Class AS, 1-Mark Bulb Pipettes offer higher measurement accuracy than graduated pipettes.
Brand: BRAND™ 230602
This item is not returnable.
View return policy
Description
- AR-Glas™
- Class 1-mark graduated
- Includes batch certificate of conformity
Specifications
Class AS | |
1-Mark Graduations | |
1 mL |
AR-GLAS | |
Batch Certificate of Conformity | |
Bulb Pipet |