Learn More
BRAND™ BLAUBRAND™ Bulb Pipettes 1-Mark
Class AS, 1-Mark Bulb Pipettes offer higher measurement accuracy than graduated pipettes.
Brand:  BRAND™ 230614
 View more versions of this product
Product Code. 16397977
  • 74.20 € / Pack of 6
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • AR-Glas™
  • Class 1-mark graduated
  • Includes batch certificate of conformity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Class AS
1-Mark Graduations
20 mL
AR-GLAS
Batch Certificate of Conformity
Bulb Pipet