Learn More
BRAND™ BLAUBRAND™ Class A Borosilicate Glass Mixing Cylinders
Includes polypropylene stopper and hexagonal base. BRAND™ Blaubrand™ Class A Borosilicate Glass Mixing Cylinders are calibrated to contain the liquid inside the vessel.
Brand:  BRAND™ 32462
124.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10757791
  • 129.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass construction (Boro 3.3. DIN EN ISO 4788)
  • Calibrated to contain (TC, In)
  • Includes polypropylene stopper and hexagonal base
  • Height without stopper
  • All BLAUBRAND™ cylinders are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer
  • On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Blue
Graduated Mixing Cylinder
DIN EN ISO 4788
1000 mL
Batch Certificate, Octagonal Polyethylene Stopper
Polyethylene
10 mL
To Contain
Round
Borosilicate Glass
B
500 mm
Blue Enamel Graduations
Polyethylene
Autoclavable