Learn More
BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Volumetric Flask with Glass Stopper is available in both narrow neck and wide neck options in a variety of sizes. DE-M marking on neck provides quick and accurate volume check.
Brand:  BRAND™ 37258
33.61 EUR valid until 2024-12-31
Use promo code "21985" to get your promotional price.
 View more versions of this product
Product Code. 11824020
  • 46.40 € / Pack of 2
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • DAkkS calibration certificate available upon request
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • DE-M marking provides quick and accurate volume check
  • Glass stopper included
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibrated To Contain
Glass
DE-M Marking
Borosilicate Glass
Narrow
Yes
10/19
DIN EN ISO 1042
Clear
25 mL
±0.04 mL
2 Pack
A