Learn More
BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Polypropylene Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flask with Polypropylene Stopper is available with narrow neck or wide neck in a variety of sizes. DE-M marking on the neck provides quick and accurate volume check. Polypropylene stopper included.
Brand:  BRAND™ 37246
26.38 EUR valid until 2025-12-31
Use promo code "24375" to get your promotional price.
 View more versions of this product
Product Code. 11854010
  • 39.20 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • DAkkS calibration certificate available upon request
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • DE-M marking provides quick and accurate volume check
  • Polypropylene stopper included
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibrated To Contain
Polypropylene
DE-M Marking
Borosilicate Glass
Narrow
Yes
Stopper
DIN EN ISO 1042
Clear
20 mL
±0.04 mL
2 Pack
A
10/19