missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BLAUBRAND™ Class AS AR-Glass Volumetric Bulb Pipet with Certificate

Calibrated to deliver (TD, Ex). BRAND™ BLAUBRAND™ Class AS AR-Glass Volumetric Bulb Pipet with USP Certificate provides a waiting time of only 5 seconds. Delivered with individual USP conformity certificate.

Brand:  BRAND™ 950616

38.35 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15353558

  • 52.75 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • AR-GLAS™ construction to DIN EN ISO 648
  • Calibrated to deliver (TD, Ex)
  • Class AS, DE-M marking
  • Waiting time 5 seconds
  • Includes USP batch certificate.
  • Delivered with individual USP conformity certificate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.03 mL USP
To Deliver
AS
Yes
Black
520 mm
Yes
DIN EN ISO 648
AR-GLAS
DE-marking
30 mL
Volumetric Bulb Pipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.