missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BlauBRAND™ Compact Burette
With PTFE stopcock, fast to dismantle and easy to clean
Brand: BRAND™ 13913
This item is not returnable.
View return policy
Description
Specifications
10 mL | |
0.02 mL | |
±0.02 mL | |
Compact Buret | |
DIN EN ISO 385 |
Borosilicate Glass | |
PTFE | |
10 mL | |
Class AS |