Learn More
Invitrogen™ BLOCK-iT™ Transfection Optimization Kit
BLOCK-iT™ Transfection Optimization Kit
Brand: Invitrogen™ 13750047
636.57 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
The BLOCK-iT™ Transfection Optimization Kit is designed to optimize RNAi transfection using controls for transfection and viability. Each kit contains a BLOCK-iT™ Fluorescent Oligo, which provides:This makes BLOCK-iT™ Fluorescent Oligo an ideal indicator of transfection efficiency for RNAi experiments. In addition, the kit contains a Positive Control Stealth RNAi™ siRNA molecule directed at human p53 and a Scrambled Stealth RNAi™ siRNA molecule as a negative control. To assay cell viability, Dead Cell Dye is included. This dye colors dead cells as red, thereby allowing easy identification of non-viable cells in a tissue culture plate. Using these reagents together allows you to both optimize transfection conditions and have a functional positive control in every RNAi experiment you perform.
Specifications
Cell Viability Control, Negative Control, Transfection Control, Positive Control | |
RNA | |
Transfection | |
Transfection Kit | |
siRNA |
Kit | |
pLATE | |
BLOCK-iT™, Stealth RNAi™ siRNA | |
1 Kit |