missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ BSA

Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.

215.00 € - 489.00 €

Specifications

Product Type Blocker BSA
Form Liquid
For Use With (Application) Western Blot, ELISA, Immunohistochemistry (IHC)
Product Line Blocker™
Concentration 10% BSA
View More Specs

Products 3
Product Code Brand Buffer Price Quantity & Availability  
Product Code Brand Buffer Price Quantity & Availability  
17992361
View Documents
Thermo Scientific™
37525X3
PBS
489.00 €
Pack of 3
Please to purchase this item. Need a web account? Register with us today!
11889310
View Documents
Thermo Scientific™
37520
TBS
215.00 €
125mL
Please to purchase this item. Need a web account? Register with us today!
10341944
View Documents
Thermo Scientific™
37525
PBS
230.00 €
200mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.

Blocker BSA is a 10% (w/v) solution of high-quality BSA that is useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. It is most frequently diluted 10-fold (to 1% BSA) in 1X PBS or 1X TBS for initial testing. Blocker BSA is usually more effective than nonfat milk for biotin-avidin systems because it contains a single purified protein that is devoid of endogenous biotin.

Specifications

Specifications

Blocker BSA
Western Blot, ELISA, Immunohistochemistry (IHC)
10% BSA
Store at 4°C.
Liquid
Blocker™
Single purified protein
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.