missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ Casein

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.

Brand:  Thermo Scientific™ 37532

117.12 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10443644

  • 192.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Features of Blocker Casein include:
Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
Easy to use—1% casein solution is ready to use; can be diluted further as needed
Safe—stable, thimerosal-free formulation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blocker Casein in TBS, 1 L
1 L
Western Blot, ELISA
1X
Single purified protein
Blocker Casein
Liquid
Blocker™
TBS
Store at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers