Learn More
Fisherbrand™ Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters
86.85 € - 109.00 €
Product Code | Brand | Diameter (Metric) | No. of Places | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | No. of Places | Price | Quantity & Availability | |||||
15327938
|
Fisherbrand™
88860101 |
6mm | 46 |
86.85 €
Each |
Estimated Shipment: 24-10-2025
Log in to see stock. |
|||||
15337938
|
Fisherbrand™
88860102 |
10 mm | 28 |
86.85 €
Each |
In Stock
Log in to see stock. |
|||||
15367938
|
Fisherbrand™
88860105 |
13 mm | 24 |
90.05 €
Each |
In Stock
Log in to see stock. |
|||||
15377938
|
Fisherbrand™
88860106 |
16 mm | 15 |
108.00 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
15387938
|
Fisherbrand™
88860107 |
18 mm | 12 |
109.00 €
Each |
In Stock
Log in to see stock. |
|||||
15397938
|
Fisherbrand™
88860108 |
20 mm | 8 |
86.85 €
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
15307948
|
Fisherbrand™
88860109 |
25 mm | 6 |
86.85 €
Each |
In Stock
Log in to see stock. |
|||||
Description
A variety of blocks for any application
- Aluminum alloy blocks
Specifications
Aluminum Alloy | |
Block |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Blocks for Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters