missing translation for 'onlineSavingsMsg'
Learn More

BD Safety-Lok™ Blood Collection Set with 30 cm Tubing

Use for blood collection. BD Vacutainer™ Safety-Lok™ Blood Collection Set with 30 cm tubing comes with safety re-shielding cover and Luer adapter.

Brand:  BD 367286

Product Code. 10247837

  • 529.00 € / Pack of 200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Includes safety re-shielding cover and Luer adapter,
  • 21 G ¾ in., 30 cm tubing, green
  • Recommended for adults
  • One case consists fo 4 x 50 Blood Collection Sets
  • One Set is necessary for a pair of blood culture bottles
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blood Collection Set
21 G
19 mm
200 Cs.
Green
With luer adapter, 21 guage, 19mm needle size, 305mm tubing
305 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.