missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 2, Sheets
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 2, Sheets are highly pure and ideal for blotting and chromatography.
40.70 € - 301.00 €
Specifications
Description | Capillary rise: 70mm/10 min., Weight: 195g/m2, 100/Box |
---|---|
Film Thickness | 0.35 mm |
Grade | BF 2 |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
15952800
|
Sartorius Stedim Biotech™
FT-2-519-080090N |
80 mm | 90 mm |
40.70 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
15962800
|
Sartorius Stedim Biotech™
FT-2-519-130210N |
130 mm | 120 mm |
96.30 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
15972800
|
Sartorius Stedim Biotech™
FT-2-519-300600N |
300 mm | 600 mm |
301.00 €
Pack of 100 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||||
Description
- This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Made of high-purity cotton linters for uniform buffer flow and resulting blots.
- No additives to avoid any interference during transfer.
- BF 2 grade is ideal for gel wicking and drying, capillary blotting using western, Southern, or semidry techniques.
Specifications
Capillary rise: 70mm/10 min., Weight: 195g/m2, 100/Box | |
0.35 mm | |
BF 2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title