missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Discs
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3 Discs are highly pure and ideal for blotting chromatography.
40.30 € - 995.00 €
Specifications
Description | Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk |
---|---|
Film Thickness | 0.76 mm |
Format | Disc |
Grade | BF 3 |
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
15922960
|
Sartorius Stedim Biotech™
FT-3-520-022 |
22 mm |
242.00 €
Pack of 1000 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
15932960
|
Sartorius Stedim Biotech™
FT-3-520-025 |
25 mm |
40.30 €
Pack of 100 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
15942960
|
Sartorius Stedim Biotech™
FT-3-520-150 |
150 mm |
995.00 €
Pack of 100 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
Description
- This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Made of high-purity cotton linters for uniform buffer flow and resulting blots.
- No additives to avoid any interference during transfer.
- BF 3 grade is ideal for increasing and maintaining the transport of liquid from the buffer and as buffer reservoir in capillary and semidry blotting methods.
Specifications
Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk | |
0.76 mm | |
Disc | |
BF 3 |