missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 4, Sheets
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 4, Sheets are highly pure and ideal for blotting chromatography.
Brand: Sartorius Stedim Biotech™ FT-2-521-110170G
This item is not returnable.
View return policy
Description
- This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Made of high-purity cotton linters for uniform buffer flow and resulting blots.
- No additives to avoid any interference during transfer.
- BF 4 grade is ideal for transferring DNA or RNA according to the Southern technique or semidry blotting of proteins.
Specifications
Capillary rise: 160mm/10 min., Weight: 550 g/m2, 100/Pk | |
1.30 mm | |
BF 4 | |
110 mm |
110 x 170 mm | |
Sheet | |
170 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction