Learn More
Buerkle™ Blue Measuring Spoon
BIOPHARMAGREENER_CHOICE
Measuring spoons in a unique shade of blue that does not occur often in foodstuffs, making them easy to find and visually identify.
Brand:  Buerkle™ 5378-4058
 View more versions of this product
Product Code. 16525250
  • 73.70 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sterile versions are individually packaged and radiation sterilized
  • The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects
  • This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry
  • Volumetric and dosing spoons can be used to measure small amounts of powder, granules and liquids exactly and dose them precisely
  • For precise measuring, excess medium is stripped off the upper edge of the spoon
  • The content volume is engraved on the handle
  • The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts
  • The flat base makes it possible to set the volumetric spoon down without spilling the content
  • The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities
  • Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded
  • The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed and the food and beverage industry
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • With stripper edge
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
Specifications
Specifications
Sampler
Dosing spoons with stripper edge for exact measuring
Single-use
Blue
Especially for The Food, Animal Feed and Pharmaceutical Industries
Polystyrene
50 mL
Non-sterile
Food Contact, Reach, RoHS
100
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number243178 Certificate TypeCertificate of Compliance Date16/08/2024 Catalog Number5378-4058
Lot Number233635 Certificate TypeCertificate of Compliance Date28/08/2023 Catalog Number5378-4058
Lot Number215482 Certificate TypeCertificate of Compliance Date18/03/2022 Catalog Number5378-4058
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Blue Measuring Spoon > Non-sterile; Capacity: 50 mL