missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Blue Measuring Spoon
BIOPHARMAGREENER_CHOICE

Measuring spoons in a unique shade of blue that does not occur often in foodstuffs, making them easy to find and visually identify.

Brand:  Buerkle™ 5378-4059

 View more versions of this product

Product Code. 16535250

  • 145.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sterile versions are individually packaged and radiation sterilized
  • The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects
  • This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry
  • Volumetric and dosing spoons can be used to measure small amounts of powder, granules and liquids exactly and dose them precisely
  • For precise measuring, excess medium is stripped off the upper edge of the spoon
  • The content volume is engraved on the handle
  • The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts
  • The flat base makes it possible to set the volumetric spoon down without spilling the content
  • The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities
  • Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded
  • The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed and the food and beverage industry
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • With stripper edge
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
Specifications

Specifications

Sampler
Dosing spoons with stripper edge for exact measuring
Single-use
Blue
Especially for The Food, Animal Feed and Pharmaceutical Industries
Polystyrene
50 mL
Sterile
Food Contact, Reach, RoHS
100
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number244328 Certificate TypeCertificate of Compliance Date07/12/2024 Catalog Number5378-4059
Lot Number243553 Certificate TypeCertificate of Compliance Date20/09/2024 Catalog Number5378-4059
Lot Number241654 Certificate TypeCertificate of Compliance Date24/05/2024 Catalog Number5378-4059
Lot Number235241 Certificate TypeCertificate of Compliance Date11/01/2024 Catalog Number5378-4059
Lot Number234137 Certificate TypeCertificate of Compliance Date13/10/2023 Catalog Number5378-4059
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ Blue Measuring Spoon > Sterile; Capacity: 50 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.