missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Blue Measuring Spoon



Measuring spoons in a unique shade of blue that does not occur often in foodstuffs, making them easy to find and visually identify.
Marca: Buerkle™ 5378-4050
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Sterile versions are individually packaged and radiation sterilized
- The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects
- This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry
- Volumetric and dosing spoons can be used to measure small amounts of powder, granules and liquids exactly and dose them precisely
- For precise measuring, excess medium is stripped off the upper edge of the spoon
- The content volume is engraved on the handle
- The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, thus allowing for good emptying of small remaining amounts
- The flat base makes it possible to set the volumetric spoon down without spilling the content
- The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities
- Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded
- The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed and the food and beverage industry
- Can be used as part of HACCP/IFS foreign object management
- Especially for the food, animal feed and pharmaceutical industries
- With stripper edge
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
Specifica
Sampler | |
Dosing spoons with stripper edge for exact measuring | |
Single-use | |
Blue | |
Especially for The Food, Animal Feed and Pharmaceutical Industries |
Polystyrene | |
5 mL | |
Non-sterile | |
Food Contact, Reach, RoHS | |
100 |
Topsellers food sampling solids
File size: 887.8 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
191118BUR5378 | Certificate of Compliance | 29/03/2023 | 5378-4050 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Buerkle™ Blue Measuring Spoon > Non-sterile; Capacity: 5 mL
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto