missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Color-coded Micro PTFE Stir Bar
Use when quick visual identification is highly important. Fisherbrand™ Color-coded Micro PTFE Stir Bar is available in three colors: blue, red, and yellow.
6.90 € - 16.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Description | PTFE encapsulated Alnico V magnet core |
Shape | Micro |
For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Color | Price | Quantity & Availability | |||
11517193
|
Fisherbrand™
001.815-B |
1.5 mm | 15 mm | Blue |
16.00 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11597183
|
Fisherbrand™
001.808-B |
1.5 mm | 8 mm | Blue |
9.10 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11567183
|
Fisherbrand™
001.802-B |
2 mm | 2 mm | Blue |
11.65 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11577183
|
Fisherbrand™
001.805-B |
2 mm | 5 mm | Blue |
11.20 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11587183
|
Fisherbrand™
001.807-B |
2 mm | 7 mm | Blue |
7.25 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
12686316
|
Fisherbrand™
001.810-B |
3 mm | 10 mm | Blue |
6.90 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11567193
|
Fisherbrand™
001.815-Y |
1.5 mm | 15 mm | Yellow |
15.60 €
Pack of 10 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
11557193
|
Fisherbrand™
001.810-Y |
3 mm | 10 mm | Yellow |
6.90 €
Pack of 10 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
11547193
|
Fisherbrand™
001.807-Y |
2 mm | 7 mm | Yellow |
7.25 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11537193
|
Fisherbrand™
001.805-Y |
2 mm | 5 mm | Yellow |
9.55 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11527193
|
Fisherbrand™
001.802-Y |
2 mm | 2 mm | Yellow |
11.65 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11537183
|
Fisherbrand™
001.808-R |
1.5 mm | 8 mm | Red |
9.10 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11557183
|
Fisherbrand™
001.815-R |
1.5 mm | 15 mm | Red |
15.60 €
Pack of 10 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
11547183
|
Fisherbrand™
001.810-R |
3 mm | 10 mm | Red |
6.90 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
11527183
|
Fisherbrand™
001.807-R |
2 mm | 7 mm | Red |
7.25 €
Pack of 10 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||
11507183
|
Fisherbrand™
001.802-R |
2 mm | 2 mm | Red |
11.65 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
12696316
|
Fisherbrand™
001.805-R |
2 mm | 5 mm | Red |
9.55 €
Pack of 10 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||
Description
- Color-coded, micro stirrer bars
- For use where identification is of prime importance
- Note: Colored PTFE coatings are not as inert as pure PTFE
Specifications
Autoclavable | |
PTFE encapsulated Alnico V magnet core | |
Containers & Vessels / Stirrer Hot Plates |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
Micro |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title