missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS 299897, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Orally active, potent γ-secretase inhibitor (IC50 = 12 nM). Inhibits Aβ40 and Aβ42 formation in vitro (IC50 values are 7.4 and 7.9 nM respectively) and reduces Aβ in the brain, plasma and cerebrospinal fluid in vivo. Exhibits no Notch toxicity. Brain penetrant.
Specifications
Specifications
CAS | 290315-45-6 |
Molecular Formula | C24H21ClF3NO4S |
Quantity | 10 mg |
Synonym | 2-1r-1-4-chlorophenyl sulfonyl 2,5-difluorophenyl amino ethyl-5-fluoro-benzenebutanoic acid, 2-1r-1-4-chlorophenyl sulfonyl 2,5-difluorophenyl amino ethyl-5-fluorobenzenebutanoic acid, 4-2-1r-1-n-2,5-difluorophenyl 4-chlorobenzenesulfonamido ethyl-5-fluorophenyl butanoic acid |
InChI Key | IZAOBRWCUGOKNH-OAHLLOKOSA-N |
SMILES | CC(C1=C(C=C(C=C1)F)CCCC(=O)O)N(C2=C(C=CC(=C2)F)F)S(=O)(=O)C3=CC=C(C=C3)Cl |
IUPAC Name | 4-[2-[(1R)-1-(N-(4-chlorophenyl)sulfonyl-2,5-difluoroanilino)ethyl]-5-fluorophenyl]butanoic acid |
Molecular Weight (g/mol) | 511.94 |
PubChem CID | 11249248 |
Formula Weight | 511.94 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction