missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS 961, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective RARγ agonist (EC50 values are 30 and 1000 nM at RARγ and RARβ respectively). Displays no activity at RARα receptors.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 399.46 |
Percent Purity | >97% |
Chemical Name or Material | BMS 961 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'