Learn More
Helios Italquartz Boats with Ring Handle, Transparent
Boats with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 62P00005
 View more versions of this product
Product Code. 15858034
  • 133.00 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transparent
  • Include ring handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Flat
Quartz
Boat with Ring Handle
76/19 mm
11 mm
Transparent
Boat
76 mm x 11 mm
11 mm