missing translation for 'onlineSavingsMsg'
Learn More

BOC-Glycine, 99+%, Thermo Scientific Chemicals

Product Code. 10066343
100 g, Glass Bottle
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4530-20-5
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.18
MDL Number MFCD00002690
InChI Key VRPJIFMKZZEXLR-UHFFFAOYSA-N
Synonym boc-glycine, boc-gly-oh, n-tert-butoxycarbonyl glycine, n-boc-glycine, n-boc-l-glycine, tert-butoxycarbonylglycine, t-butoxycarbonylglycine, 2-tert-butoxycarbonyl amino acetic acid, glycine, n-1,1-dimethylethoxy carbonyl, tert-butyloxycarbonylglycine
PubChem CID 78288
SMILES CC(C)(C)OC(=O)NCC(O)=O

Specifications

Melting Point 87°C to 90°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass Bottle
Linear Formula (CH3)3COCONHCH2CO2H
Beilstein 04, 2411
Solubility Information Solubility in water: soluble in water
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
Quantity 100 g
Formula Weight 175.18
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material BOC-Glycine
Show More Show Less
compliance-icons
Product Identifier
  • BOC-Glycine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BOC-Glycine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.