missing translation for 'onlineSavingsMsg'
Learn More
BOC-L-Valine, 99+%, Thermo Scientific Chemicals Product Code.: 10145503

BOC-L-Valine, 99+%, Thermo Scientific Chemicals

Product Code. 10145503
100 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10145503

Brand: Thermo Scientific Acros 275711000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13734-41-3
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00065605
InChI Key SZXBQTSZISFIAO-ZETCQYMHSA-N
Synonym boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine
PubChem CID 83693
SMILES CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 76°C to 80°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula (CH3)2CHCHCOOHNHCOOC(CH3)3
Specific Rotation -5.3° to -7.3° (20°C, 589nm) (c=1, CH3COOH)
IUPAC Name (2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Specific Rotation Condition - 6.30 (20.00°C c=1,CH3COOH)
Quantity 100 g
Formula Weight 217.27
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material BOC-L-Valine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463408 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number275711000,275710050,275710250
Lot NumberA0380529 Certificate TypeCertificate of Analysis Date31/01/2022 Catalog Number275711000,275710050,275710250
Lot NumberA0437329 Certificate TypeCertificate of Analysis Date23/12/2021 Catalog Number275711000,275710050,275710250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BOC-L-Valine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.