missing translation for 'onlineSavingsMsg'
Learn More

BOC-ON, 99%, Thermo Scientific Chemicals

Product Code. 10594644
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58632-95-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00001863
InChI Key QQWYQAQQADNEIC-PTNGSMBKSA-N
Synonym 2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino
PubChem CID 5868400
IUPAC Name tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate
SMILES CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1

Specifications

Melting Point 86°C to 89°C
Color White to Beige
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3COCO2N=C(C6H5)CN
Quantity 100 g
Fieser 06,91; 10,61
Merck Index 15, 1323
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethyl acetate,ether,benzene,dioxane,,chloroform,acetone,methanol,isopropanol and,tert-butanol,insoluble in petroleum ether
Formula Weight 246.27
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material BOC-ON, 99%
Show More Show Less
compliance-icons
Product Identifier
  • BOC-ON
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'