missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ BOD Control Test Tablet

Velp Scientifica™ BOD Measurement Test Tablet for use with BOD Sensors and BOD EVO Sensors.

Brand:  Velp Scientifica™ A00000136

59.84 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.



Product Code. 11771864

  • 70.15 € / Pack of 10
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with BOD and BOD EVO Sensors
Specifications

Specifications

8 Pack
BOD Control Test Tablet
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : boric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers