Learn More
WTW™ BOD Test Kits
Obtain precision photometric measurements for waste water, drinking water, and surface water with WTW™ BOD test Kits.
Brand:  WTW™ 252028
Product Code. 15381437
  • 160.00 € / 1 set
Explore more special offers
Description
Description
  • For wastewater, production control, ground water, surface water
  • For use with PhotoLab™ models S6, S12 and 6000/7000
  • Biological Oxygen Demand (BOD) Test Kits
  • This test is highly accurate and easy to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PhotoLab™ Photometers
50
0.5 to 3000 mg/L BOD
Photometric