missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ BODIPY™ TMR C5-Maleimide
BODIPY™ TMR C5-maleimide
Brand: Molecular Probes™ B30466
216.01 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The thiol-reactive BODIPY™ TMR maleimide produces electronically neutral dye conjugates that are spectrally similar to the positively charged tetramethylrhodamine dye. This dye's lack of ionic charge results in minimal effects on the isoelectric points of standard proteins conjugated with this fluorophore. The small size and relatively long excited-state lifetime of BODIPY™ TMR dye has proven useful for studying ligand-receptor interaction by fluorescence polarization.Specifications
1 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
BODIPY™ TMR | |
BODIPY™ |
Maleimide | |
Thiol | |
Maleimide | |
BODIPY Dyes |
For Research Use Only. Not for use in diagnostic procedures.