Learn More
Grant Instruments™ Test Tube Rack for Unstirred Water Baths
Keeps test tubes upright and secure in the bath.
Brand:  Grant Instruments™ J2-LE
 View more versions of this product
Product Code. 11476397
  • 146.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Keeps test tubes upright and secure in the bath.
  • Compatible with a wide range of Grant Unstirred Water Baths
  • Holds up to 105 test tubes depending on size
  • Dual slotted trays keep tubes secure and prevent tipping
Specifications
Specifications
65 1.5 mL Tubes