missing translation for 'onlineSavingsMsg'
Learn More

Boiling Stones, Honeywell Fluka™

sieve analysis > 1 mm, >95%

Brand:  Honeywell-Fluka 85106-500G

Product Code. 15637340

  • 159.74 € / 500g
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Boiling Stones
500 g
2980°C
Poly Bottle
2038°C
9.4 to 10.1 (at 20°C)
1344-28-1
NONH for all modes of transport
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.