Learn More
Bohlender™ BOLA™ Connectors for Sampling Bags
PTFE connectors for sampling bags.
Brand:  Bohlender™ A226-06
Product Code. 17126934
  • 24.00 € / Pack of 2
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Connectors to be screwed in the ports of sampling bags
  • Bore diameter: 3 mm
Specifications
Specifications
PTFE
0.24 in.
6 mm